2,4-Dimethylthiophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Mercapto-m-xylene1,3-Dimethyl-4-mercaptobenzene2,4-Dimethylbenzene-1-thiol2,4-Dimethylbenzenethiol

OR7946-25G OR7946-5G OR7946-100G
76612-436EA 17 USD
76612-436 76612-434 76612-432
2,4-Dimethylthiophenol 95%
2,4-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 207…208 °C
Density: 1.022 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010019
CAS Number: 13616-82-5
EINECS: 237-100-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR