4-Benzyloxy-3-methoxyphenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 2-(4-(Benzyloxy)-3-methoxyphenyl)acetonitrile

OR7895-250MG
76625-456EA 15 USD
76625-456
4-Benzyloxy-3-methoxyphenylacetonitrile 98%
4-Benzyloxy-3-methoxyphenylacetonitrile
Formula: C₁₆H₁₅NO₂
MW: 253.30 g/mol
Boiling Pt: 195 °C/1mmHg
Melting Pt: 68…70 °C
Storage Temperature: Ambient
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR