2-Amino-5-chloro-3-iodobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

OR70215-1G OR70215-5G
76625-434EA 389 USD
76625-434 76625-432
2-Amino-5-chloro-3-iodobenzonitrile
2-Amino-5-chloro-3-iodobenzonitrile
Formula: C₇H₄ClIN₂
Storage Temperature: Ambient
MDL Number: MFCD09800802
CAS Number: 1000577-48-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR