2,5-Dibromo-p-xylene 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene1,4-Dibromo-2,5-dimethylbenzene

OR70071-100G OR70071-25G OR70071-500G
76617-282EA 16 USD
76617-282 76617-284 76617-280
2,5-Dibromo-p-xylene 95%
2,5-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 261 °C
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR