2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR6739-250MG
76595-106EA 42 USD
76595-106
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde
Formula: C₉H₈N₂O
MW: 160.1700 g/mol
Melting Pt: 117…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02251327
CAS Number: 30384-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR