2,6-Dichloro-3-iodopyridine 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR6713-1G OR6713-5G
76617-240EA 13.39 USD
76617-240 76617-242
2,6-Dichloro-3-iodopyridine 95%
2,6-Dichloro-3-iodopyridine
Formula: C₅H₂Cl₂IN
MW: 273.88 g/mol
Melting Pt: 74…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094686
CAS Number: 148493-37-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR