α-Chloro-2-methyl-3-nitrotoluene

Supplier: APOLLO SCIENTIFIC

Danger

OR6708-5G OR6708-25G OR6708-100G
76621-820EA 492 USD
76621-820 76621-822 76621-824
α-Chloro-2-methyl-3-nitrotoluene
α-Chloro-2-methyl-3-nitrotoluene
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 43…46 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007165
CAS Number: 60468-54-4
EINECS: 262-249-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR