5-Bromo-2,4-dimethoxypyrimidine

Supplier: APOLLO SCIENTIFIC

Warning

OR6687-25G OR6687-1G OR6687-5G
76595-050EA 37 USD
76595-050 76595-046 76595-048
5-Bromo-2,4-dimethoxypyrimidine
5-Bromo-2,4-dimethoxypyrimidine
Formula: C₆H₇BrN₂O₂
MW: 219.0400 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00038016
CAS Number: 56686-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR