4-(Phenylsulfanyl)aniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Phenylthio)aniline

OR6679-1G OR6679-5G
76595-032EA 42 USD
76595-032 76595-034
4-(Phenylsulfanyl)aniline
4-(Phenylsulfanyl)aniline
Formula: C₁₂H₁₁NS
MW: 201.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00094027
CAS Number: 1135-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR