Cyclopentylacetic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Cyclopentylacetic acid

OR6624-25G OR6624-5G OR6624-100G
76594-992EA 98 USD
76594-992 76594-990 76594-994
Cyclopentylacetic acid
Cyclopentylacetic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 133…134 °C/23mmHg
Density: 1.023 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001387
CAS Number: 1123-00-8
EINECS: 214-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR