Diethyl mesoxalate (diethyl ketomalonate) 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Diethyl 2-oxomalonate

OR6568-100G OR6568-25G OR6568-5G
76594-936EA 113 USD
76594-936 76594-934 76594-932
Diethyl mesoxalate (diethyl ketomalonate) 97%
Diethyl mesoxalate (Diethyl ketomalonate)
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 208…210 °C
Density: 1.142 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
EINECS: 210-176-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR