1-Indanone 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2,3-Dihydro-1H-inden-1-one

OR6561-100G OR6561-25G OR6561-10G
76594-920EA 18 USD
76594-920 76594-924 76594-922
1-Indanone 98%
1-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 243…245 °C
Melting Pt: 38…42 °C
Density: 1.103 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003785
CAS Number: 83-33-0
EINECS: 201-470-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR