Aminoacetaldehyde diethyl acetal

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,2-Diethoxyethanamine2-Aminoacetaldehyde diethylacetal

OR6496-5G
76612-280EA 13.39 USD
76612-280
Aminoacetaldehyde diethyl acetal
Aminoacetaldehyde diethyl acetal
Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Boiling Pt: 161…163 °C
Melting Pt: –78 °C
Density: 0.916 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008136
CAS Number: 645-36-3
EINECS: 211-439-4
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR