2,6-Pyridinedimethanol

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,6-Bis(hydroxymethyl)pyridine

OR6456-100G OR6456-5G OR6456-25G
76594-794EA 63 USD
76594-794 76594-792 76594-796
2,6-Pyridinedimethanol
2,6-Pyridinedimethanol
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 185 °C/15mmHg
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00006351
CAS Number: 1195-59-1
EINECS: 214-803-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR