1H-Benzimidazole-4-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H-Benzimidazole-7-carboxylic acid

OR6307-1G OR6307-250MG
76617-190EA 51 USD
76617-190 76617-192
1H-Benzimidazole-4-carboxylic acid 97%
1H-Benzimidazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
Melting Pt: 326…330 °C
Storage Temperature: Ambient
MDL Number: MFCD01823426
CAS Number: 46006-36-4
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR