Methyl-4-ethynylbenzoate 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 4-ethynylbenzoate
OR63024-25G
OR63024-5G
OR63024-1G
76594-662EA
31
USD
76594-662
76594-666
76594-664
Methyl-4-ethynylbenzoate 98%
Methyl 4-ethynylbenzoate
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Melting Pt: 92…95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00168820 CAS Number: 3034-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...