5-(4-Chlorophenyl)-4-oxazolecarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR6278-1G OR6278-100MG
76594-626EA 291 USD
76594-626 76594-624
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid 98%
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid
Formula: C₁₀H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00105450
CAS Number: 143659-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR