2-Bromo-3-nitroanisole 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-1-methoxy-3-nitrobenzene

OR6218-25G OR6218-1G OR6218-5G
76594-552EA 13.39 USD
76594-552 76594-556 76594-554
2-Bromo-3-nitroanisole 99%
2-Bromo-3-nitroanisole
Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
Melting Pt: 93…104 °C
Storage Temperature: Ambient
MDL Number: MFCD07779272
CAS Number: 67853-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR