2-Chloro-5-formylpyridine 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-Chloronicotinaldehyde6-Chloropyridine-3-carboxaldehyde2-Chloropyridine-5-carboxaldehyde

OR6178-5G OR6178-1G
76594-420EA 13 USD
76594-420 76594-422
2-Chloro-5-formylpyridine 95%
2-Chloro-5-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03095223
CAS Number: 23100-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR