4-Iodobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR61519-1G OR61519-5G OR61519-25G
76594-344EA 26 USD
76594-344 76594-342 76594-346
4-Iodobenzaldehyde 98%
4-Iodobenzaldehyde
Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 265 °C
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00039576
CAS Number: 15164-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR