3-Tolualdehyde 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: m-Tolylaldehydem-Tolualdehyde3-Methylbenzaldehyde

OR61503-500G OR61503-100G OR61503-1KG
76594-340EA 264 USD
76594-340 76594-336 76594-338
3-Tolualdehyde 98%
3-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 198…200 °C
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 204-617-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR