2-Bromo-p-anisidine 96%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Bromo-4-methoxyaniline

OR61502-1G OR61502-5G OR61502-25G
76617-160EA 47 USD
76617-160 76617-158 76617-156
2-Bromo-p-anisidine 96%
2-Bromo-p-anisidine
Formula: C₇H₈BrNO
MW: 202.0485 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09029057
CAS Number: 32338-02-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR