3-Bromo-4-iodoanisole 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Bromo-1-iodo-4-methoxybenzene

OR61448-5G OR61448-25G OR61448-1G
76594-322EA 130 USD
76594-322 76594-320 76594-324
3-Bromo-4-iodoanisole 97%
3-Bromo-4-iodoanisole
Formula: C₇H₆BrIO
MW: 312.9304 g/mol
Boiling Pt: 90 °C/2mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD12547848
CAS Number: 466639-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR