3-Methyl-2-nitrobenzoic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Methyl-2-nitrobenzoic acid2-Nitro-m-toluic acid

OR61446-25G
76594-318EA 13 USD
76594-318
3-Methyl-2-nitrobenzoic acid
2-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00007180
CAS Number: 5437-38-7
EINECS: 226-610-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR