2-Amino-3-chloropyrazine 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Chloropyrazin-2-amine3-Chloro-2-pyrazinamine

OR6127-25G OR6127-5G OR6127-1G
76594-168EA 18 USD
76594-168 76594-166 76594-170
2-Amino-3-chloropyrazine 97%
2-Amino-3-chloropyrazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Boiling Pt: 248 °C
Melting Pt: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD04114305
CAS Number: 6863-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR