4-Tolualdehyde 96%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Methylbenzaldehydp-Tolualdehyde4-Tolylaldehydep-Methylbenzaldehyd4-Methylbenzaldehydep-Tolylaldehyde

OR61250-100G OR61250-500G OR61250-2.5KG
76594-152EA 52 USD
76594-152 76594-150 76594-154
4-Tolualdehyde 96%
4-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 204 °C
Melting Pt: –6 °C
Density: 1.018 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR