3,5-Xylidine 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 5-Amino-m-xylene3,5-Dimethylaniline

OR61129-100G OR61129-25G OR61129-5G
76612-224EA 20 USD
76612-224 76612-222 76612-220
3,5-Xylidine 97%
3,5-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 221 °C
Melting Pt: 44446 °C
Density: 0.972 g/cm³ (25 °C)
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR