2-Amino-5-chloropyrazine

Supplier: APOLLO SCIENTIFIC
Warning

OR61102-5G OR61102-1G
76594-090EA 38 USD
76594-090 76594-092
2-Amino-5-chloropyrazine
2-Amino-5-chloropyrazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03423596
CAS Number: 33332-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR