4-Methyl-3-pyridineboronic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-Methylpyridin-3-yl)boronic acid4-Methylpyridine-3-boronic acid

OR6103-1G OR6103-5G
76593-980EA 33 USD
76593-980 76593-982
4-Methyl-3-pyridineboronic acid 96%
4-Methyl-3-pyridineboronic acid
Formula: C₆H₈BNO₂
MW: 136.94 g/mol
Melting Pt: 102…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788244
CAS Number: 148546-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR