6-Isoquinolinol 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-HydroxyisoquinolineIsoquinolin-6-ol

OR6099-500MG OR6099-1G OR6099-250MG
76593-956EA 43 USD
76593-956 76593-952 76593-954
6-Isoquinolinol 98%
6-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114860
CAS Number: 7651-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR