1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: O-(2-Chlorobenzyl)hydroxylamine hydrochloride

OR6059-250MG OR6059-1G
76593-930EA 31 USD
76593-930 76593-932
1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride
1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride
Formula: C₇H₉Cl₂NO
MW: 194.0585 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD01114592
CAS Number: 5555-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR