2-Ethyl-5-nitrobenzoxazole

Supplier: APOLLO SCIENTIFIC
Warning

OR60171-1G
76593-788EA 47 USD
76593-788
2-Ethyl-5-nitrobenzoxazole
2-Ethyl-5-nitrobenzoxazole
Formula: C₉H₈N₂O₃
Storage Temperature: Ambient
CAS Number: 204771-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR