2-Formyl-3-picoline 96%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Methylpyridine-2-carboxaldehyde3-Methylpicolinaldehyde

OR60091-5G OR60091-1G
76593-670EA 69 USD
76593-670 76593-668
2-Formyl-3-picoline 96%
2-Formyl-3-picoline
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 84 °C/12.8mmHg
Melting Pt: 8 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD02181144
CAS Number: 55589-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR