1H-Indazole-7-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

OR60064-10G OR60064-1G OR60064-5G
76593-636EA 63 USD
76593-636 76593-638 76593-634
1H-Indazole-7-carboxylic acid 97%
1H-Indazole-7-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD06804572
CAS Number: 677304-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR