1H-Indazole-6-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC

Warning

OR60063-1G
76593-632EA 19 USD
76593-632
1H-Indazole-6-carboxylic acid 95%
1H-Indazole-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
Melting Pt: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD06804571
CAS Number: 704-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR