1H-Indazole-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR60062-5G OR60062-1G
76593-630EA 68 USD
76593-630 76593-628
1H-Indazole-5-carboxylic acid
1H-Indazole-5-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 318…322 °C
Storage Temperature: Ambient
MDL Number: MFCD06804570
CAS Number: 61700-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR