2-Amino-2-(3-bromophenyl)acetic acid 95+%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-DL-phenylglycine

OR60052-250MG
76593-626EA 18 USD
76593-626
2-Amino-2-(3-bromophenyl)acetic acid 95+%
2-Amino-2-(3-bromophenyl)acetic acid
Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665358
CAS Number: 79422-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR