2,6-Dichloronicotinonitrile 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 2,6-Dichloro-3-cyanopyridine2,6-Dichloropyridine-3-carbonitrile

OR60049-5G OR60049-1G OR60049-25G
76617-104EA 132 USD
76617-104 76617-106 76617-102
2,6-Dichloronicotinonitrile 97%
2,6-Dichloronicotinonitrile
Formula: C₆H₂Cl₂N₂
MW: 173.00 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR