4-Indoleboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR60010-10G OR60010-5G OR60010-1G
76593-592EA 129 USD
76593-592 76593-594 76593-590
4-Indoleboronic acid pinacol ester
4-Indoleboronic acid pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
Melting Pt: 168…173 °C
Storage Temperature: Ambient
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR