2-Bromo-5-methoxyaniline 97%

Supplier: APOLLO SCIENTIFIC

Warning

OR59916-5G OR59916-100G OR59916-25G
76593-530EA 47 USD
76593-530 76593-532 76593-528
2-Bromo-5-methoxyaniline 97%
2-Bromo-5-methoxyaniline
Formula: C₇H₈BrNO
MW: 202.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070741
CAS Number: 59557-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR