2-Bromofluorene

Supplier: APOLLO SCIENTIFIC
Warning

OR5981-100G OR5981-25G OR5981-5G
76593-382EA 14.42 USD
76593-382 76593-384 76593-380
2-Bromofluorene
2-Bromofluorene
Formula: C₁₃H₉Br
MW: 245.12 g/mol
Boiling Pt: 185 °C/135mmHg
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00001115
CAS Number: 1133-80-8
EINECS: 214-480-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR