1-Acetylpiperidine-4-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR5959-100G OR5959-25G
76593-364EA 92 USD
76593-364 76593-362
1-Acetylpiperidine-4-carboxylic acid
1-Acetylpiperidine-4-carboxylic acid
Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00023706
CAS Number: 25503-90-6
EINECS: 247-049-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR