1-Bromoisoquinoline

Supplier: APOLLO SCIENTIFIC

Warning

OR59454-1G
76617-070EA 80 USD
76617-070
1-Bromoisoquinoline
1-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.0546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234478
CAS Number: 1532-71-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR