Imidazo[1,2-a]pyridine-7-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR59396-1G OR59396-5G
76593-276EA 99 USD
76593-276 76593-274
Imidazo[1,2-a]pyridine-7-carboxylic acid
Imidazo[1,2-a]pyridine-7-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
Melting Pt: 244…247 °C
Storage Temperature: Ambient
MDL Number: MFCD09834781
CAS Number: 648423-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR