4-Phenylbenzoic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: [1,1'-Biphenyl]-4-carboxylic acidBiphenyl-4-carboxylic acid4-Biphenylcarboxylic acid

OR5838-500G OR5838-100G
76593-112EA 27 USD
76593-112 76593-114
4-Phenylbenzoic acid 98%
4-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 224…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00002553
CAS Number: 92-92-2
EINECS: 202-203-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR