2-Bromo-6-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC

Danger

OR5747-100G OR5747-25G
76593-010EA 464 USD
76593-010 76593-008
2-Bromo-6-chloropyridine 98%
2-Bromo-6-chloropyridine
Formula: C₅H₃BrClN
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00181262
CAS Number: 5140-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR