5-(Boc-amino)-2-bromopyridine 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5-Amino-2-bromopyridine, 5-BOC protectedtert-Butyl (6-bromopyridin-3-yl)carbamate

OR5734-1G OR5734-250MG
76592-970EA 25 USD
76592-970 76592-968
5-(Boc-amino)-2-bromopyridine 95%
5-(Boc-amino)-2-bromopyridine
Formula: C₁₀H₁₃BrN₂O₂
MW: 273.13 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD07363808
CAS Number: 218594-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR