2-Amino-4-thiazolecarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Aminothiazole-4-carboxylic acid

OR5587-100G OR5587-25G
76592-808EA 182 USD
76592-808 76592-806
2-Amino-4-thiazolecarboxylic acid 98%
2-Amino-4-thiazolecarboxylic acid
Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
Melting Pt: 245…247 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00859429
CAS Number: 40283-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR