5-Bromo-4-chloro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR55605-5G OR55605-250MG OR55605-1G
76592-598EA 43 USD
76592-598 76592-596 76592-600
5-Bromo-4-chloro-1H-indazole
5-Bromo-4-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11007933
CAS Number: 1082041-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR