Di-tert-butyl malonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR55346-100G OR55346-500G OR55346-25G
76592-442EA 240 USD
76592-442 76592-440 76592-438
Di-tert-butyl malonate 98%
Di-tert-butyl malonate
Formula: C₁₁H₂₀O₄
Boiling Pt: 65…67 °C/1mmHg
Density: 1.42 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00008810
CAS Number: 541-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR